Esta buscando: Tests de microorganismos
465  results were found
SearchResultCount:"465"
Sort Results
Vista lista Vista Extendida (Nueva)
Valore los resultados de su búsqueda
Proveedor: VWR Chemicals
Descripción: Microorganism Tests, VWR®, Prueba de coagulasa, 3 ml
Proveedor: VWR Chemicals
Descripción: Microorganism Tests, VWR®, Cryoinstant, amarillo
MSDS Certificados Promoción
Numero del catalogo: (OXOIR4607024)
Proveedor: THERMO FISHER DIAGNOSTICS GMBH
Descripción: Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the ...
UOM: 1 * 5 UN
Numero del catalogo: (OXOIR4607049)
Proveedor: THERMO FISHER DIAGNOSTICS GMBH
Descripción: Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification...
UOM: 1 * 5 UN
Numero del catalogo: (OXOIR4606512)
Proveedor: THERMO FISHER DIAGNOSTICS GMBH
Descripción: Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the ...
UOM: 1 * 5 UN
Numero del catalogo: (OXOIR4602050)
Proveedor: THERMO FISHER DIAGNOSTICS GMBH
Descripción: Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the ...
UOM: 1 * 5 UN
Numero del catalogo: (OXOIR4601544)
Proveedor: THERMO FISHER DIAGNOSTICS GMBH
Descripción: Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the ...
UOM: 1 * 5 UN
New Product
Proveedor: LIOFILCHEM S.R.L.
Descripción: Quanti-CultiControl are lyophilized, quantitative microorganism preparations to be used in industrial laboratories for Quality Control purposes. Proce...
Numero del catalogo: (OXOIR4607090)
Proveedor: THERMO FISHER DIAGNOSTICS GMBH
Descripción: Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the ...
UOM: 1 * 5 UN
New Product
Numero del catalogo: (OXOIR4609013)
Proveedor: THERMO FISHER DIAGNOSTICS GMBH
Descripción: Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the ...
UOM: 1 * 5 UN
New Product
Numero del catalogo: (OXOIR4601241)
Proveedor: THERMO FISHER DIAGNOSTICS GMBH
Descripción: Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the ...
UOM: 1 * 5 UN
New Product
Numero del catalogo: (OXOIR4606502)
Proveedor: THERMO FISHER DIAGNOSTICS GMBH
Descripción: Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the ...
UOM: 1 * 5 UN
New Product
Numero del catalogo: (OXOIR4604000)
Proveedor: THERMO FISHER DIAGNOSTICS GMBH
Descripción: Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the ...
UOM: 1 * 5 UN
New Product
Numero del catalogo: (OXOIR4601906)
Proveedor: THERMO FISHER DIAGNOSTICS GMBH
Descripción: Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the ...
UOM: 1 * 5 UN
New Product
Numero del catalogo: (OXOIR4601996)
Proveedor: THERMO FISHER DIAGNOSTICS GMBH
Descripción: Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the ...
UOM: 1 * 5 UN
New Product
Numero del catalogo: (OXOIR4604010)
Proveedor: THERMO FISHER DIAGNOSTICS GMBH
Descripción: Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the ...
UOM: 1 * 5 UN
New Product
Consulta de precio
El stock para este material es limitada pero puede estar disponible en un almacén cerca de usted. Por favor, asegúrese de que ha iniciado sesión en la web para que el stock disponible se puede mostrar. Si el call sigue apareciendo y usted necesita ayuda, por favor llámenos al 902 222 897 o por email en webshop.es@avantorsciences.com.
El stock para este material es limitada pero puede estar disponible en un almacén cerca de usted. Por favor, asegúrese de que ha iniciado sesión en la web para que el stock disponible se puede mostrar. Si el call sigue apareciendo y usted necesita ayuda, por favor llámenos al 902 222 897 o por email en webshop.es@avantorsciences.com.
Este producto se trata de un artículo regulado sometido a normativa que restringe su venta. Si procede, nos pondremos en contacto con usted para solicitarle la licencia o declaración de uso necesaria para poder proceder al suministro del producto.
Este producto se trata de un artículo regulado sometido a normativa que restringe su venta.
Si procede, nos pondremos en contacto con usted para solicitarle la licencia o declaración de uso necesaria para poder proceder al suministro del producto.
Este producto ha sido bloqueado por su organización. Por favor, pónganse en contacto con su departamento de compras para obtener más información.
El producto original ya no está disponible. Su sustituto se muestra a continuación.
El producto(s) marcados con este símbolo están descatalogados - se venden hasta acabar stock. Pueden encontrar alternativas buscando con el número de catálogo VWR listado arriba. Si necesita más ayuda, por favor llame al Servicio de atención al cliente de VWR al 902.222.897.
1 - 16 of 465
no targeter for Bottom