Ácido 2-hidroxinicotinico ≥98.0% (by GC, titration analysis)
Proveedor: TCI EUROPE
|
Atención
|
Sinónimos:
Ácido 1,2-dihidro-2-oxonicotínico
TCIAH0795-25GEA
80
EUR
TCIAH0795-25G
Ácido 2-hidroxinicotinico ≥98.0% (by GC, titration analysis)
Ácido 2-hidroxinicotinico
Formula:
C₆H₅NO₃ MW: 139,11 g/mol |
Número MDL:
MFCD00010100 Núm. CAS: 609-71-2 EINECS: 210-198-2 |
![]() |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystaline |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in hot Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...