trans-2,3-Dimethoxycinnamic acid ≥98.0% (by titrimetric analysis)
Proveedor: TCI EUROPE
|
Atención
|
TCIAD2326-25GEA
139
EUR
TCIAD2326-25G
trans-2,3-Dimethoxycinnamic acid ≥98.0% (by titrimetric analysis)
trans-2,3-Dimethoxycinnamic acid
Formula:
C₁₁H₁₂O₄ MW: 208,21 |
Número MDL:
MFCD00004376 Núm. CAS: 7345-82-6 |
![]() |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 180.0 to 184.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...