Semi-dry transfer units, TE70XP and TE77XP
Proveedor: Hoefer
732-2189EA 2154 EUR
HOEFTE70XP HOEFTE77XP
Semi-dry transfer units, TE70XP and TE77XP
The TE70XP and TE77XP Semi-Dry Transfer Units include an intelligent built-in power supply, which automatically monitors the transfer status, stopping the transfer before the stack overheats.
  • Intelligent built-in power supply prevents the stack from overheating by monitoring the transfer status
  • Requires minimal current so does not generate the excessive heat that can dry out the transfer stack and halt transfer or damage transfer units
  • Durable iridium oxide and stainless steel electrodes allow for contamination-free, consistent transfer
  • Vented electrodes prevent build-up of bubbles which may impair transfer
  • Minimal buffer requirements reduce reagent cost and preparation time
Información de suministro: Both systems include moulded base with platinum coated titanium anode and internal power supply, hinged lid with stainless steel cathode, and 50× porous cellophane sheets (20×35,5 cm). In addition, TE70XP includes 25× blotting paper sheets (14×16 cm) and 2× Mylar® masks  (16,5×18,5 cm). TE77XP includes 25× blotting paper sheets (21×26 cm) and 2× Mylar® masks (23×27,5 cm).