Slot blot blotting manifold, PR648
Proveedor: Hoefer
HOEFPR648EA 1194 EUR
HOEFPR648
Slot blot blotting manifold, PR648
Información de suministro: Each manifold includes top block, membrane support block with O-ring, bottom block with connector, QuickFit connector with O-ring, 6× clamping screws, and membrane cutting template.