Indometacina

PROOICRS0330EA 0 EUR
PROOICRS0330
Indometacina
Indometacina
Formula: C₁₉H₁₆ClNO₄
MW: 357,80 g/mol
Número MDL: MFCD00057095
Núm. CAS: 53-86-1
EINECS: 200-186-5
UN: 2811
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR