Sudan black B ≥99.0%, grado alta pureza

Proveedor: VWR Chemicals

0593-50GEA 178 EUR
0593-50G 0593-25G
Sudan black B ≥99.0%, grado alta pureza
Sudan black B

A lipid stain used for demonstrating triglycerides in frozen sections. Also a histochemical stain for chromosomes, golgi apparatus, and leucocyte granules.

Formula: C₂₉H₂₄N₆
MW: 456,55 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00006919
Núm. CAS: 4197-25-5
EINECS: 224-087-1

Order Now

Resultados test especificaciones

Lambda max 596 – 605 nm
Melting point 118 – 126 °C
P–15/P+15 0.98 – 1.02

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR