Esta buscando: Anticaídas
27  results were found
SearchResultCount:"27"
Sort Results
Vista lista Vista Extendida (Nueva)
Valore los resultados de su búsqueda
Numero del catalogo: (MMMA3504426)
Proveedor: 3M
Descripción: Anchorage connectors are built from top quality materials guaranteeing high strength and durability to stand up to the toughest environments. Anchorag...
UOM: 1 * 1 UN
New Product
Proveedor: CAPITAL SAFETY DEUTSCHLAND
Descripción: These Protecta® Rebel™ Web self-retracting lifelines provide an economical fall protection solution without compromising on performance or safety. The...
Numero del catalogo: (CPSD1500089)
Proveedor: CAPITAL SAFETY DEUTSCHLAND
Descripción: The DBI-SALA® adjustable radio and mobile phone holster is specially designed to adjust perfectly to the size of virtually any portable radio, as well...
UOM: 1 * 1 UN
Proveedor: CAPITAL SAFETY DEUTSCHLAND
Descripción: This pole hoist attaches to a carabiner on the universal bracket to a suitable anchor point, allowing to work in any direction from any angle.
Numero del catalogo: (CPSD1500009)
Proveedor: CAPITAL SAFETY DEUTSCHLAND
Descripción: D-Ring cord attachment point, 2,3 kg (5 lbs.) capacity.
UOM: 1 * 10 UN
Proveedor: CAPITAL SAFETY DEUTSCHLAND
Descripción: Sealed-Blok™ family has the most comprehensive line of sealed SRLs in the industry and decades of proven field service in the toughest environments.
Numero del catalogo: (CPSD1500109)
Proveedor: CAPITAL SAFETY DEUTSCHLAND
Descripción: Dual tool harness holster with two retractors.
UOM: 1 * 1 UN
Proveedor: CAPITAL SAFETY DEUTSCHLAND
Descripción: Tripod with adjustable locking legs, safety chains, top pulley and winch bracket.
Numero del catalogo: (CPSD2104565)
Proveedor: CAPITAL SAFETY DEUTSCHLAND
Descripción: Reusable D-Ring assembly in a rugged design for harsh environments.
UOM: 1 * 1 UN
Numero del catalogo: (CPSD1500104)
Proveedor: CAPITAL SAFETY DEUTSCHLAND
Descripción: Single tool harness holster with retractor.
UOM: 1 * 1 UN
Numero del catalogo: (CPSD1500003)
Proveedor: CAPITAL SAFETY DEUTSCHLAND
Descripción: D-rings can be attached to virtually any tool under 2,3 kg (5 lbs.) or 0,9 kg (2 lbs.) by using Quick-Wrap Tape.
UOM: 1 * 1 UN
Numero del catalogo: (CPSDAM211)
Proveedor: CAPITAL SAFETY DEUTSCHLAND
Descripción: Protecta® anchorage D-ring made from 316 stainless steel; 12 mm fixing.
UOM: 1 * 1 UN
Numero del catalogo: (CPSDAM210)
Proveedor: CAPITAL SAFETY DEUTSCHLAND
Descripción: Protecta® Fixed anchorage plate made from stainless steel.
UOM: 1 * 1 UN
Numero del catalogo: (CPSD1500092)
Proveedor: CAPITAL SAFETY DEUTSCHLAND
Descripción: Spray can/water bottle holster with coil tether.
UOM: 1 * 1 UN
Numero del catalogo: (CPSD2108413)
Proveedor: CAPITAL SAFETY DEUTSCHLAND
Descripción: The DBI-SALA® fixed beam anchor fits 63 to 457 mm wide i-beams up to 38 mm thick. It installs in seconds to the beam structure in any orientation with...
UOM: 1 * 1 UN
Numero del catalogo: (CPSD1500099)
Proveedor: CAPITAL SAFETY DEUTSCHLAND
Descripción: Tape measure holster.
UOM: 1 * 1 UN
Consulta de precio
El stock para este material es limitada pero puede estar disponible en un almacén cerca de usted. Por favor, asegúrese de que ha iniciado sesión en la web para que el stock disponible se puede mostrar. Si el call sigue apareciendo y usted necesita ayuda, por favor llámenos al 902 222 897 o por email en webshop.es@avantorsciences.com.
El stock para este material es limitada pero puede estar disponible en un almacén cerca de usted. Por favor, asegúrese de que ha iniciado sesión en la web para que el stock disponible se puede mostrar. Si el call sigue apareciendo y usted necesita ayuda, por favor llámenos al 902 222 897 o por email en webshop.es@avantorsciences.com.
Este producto se trata de un artículo regulado sometido a normativa que restringe su venta. Si procede, nos pondremos en contacto con usted para solicitarle la licencia o declaración de uso necesaria para poder proceder al suministro del producto.
Este producto se trata de un artículo regulado sometido a normativa que restringe su venta.
Si procede, nos pondremos en contacto con usted para solicitarle la licencia o declaración de uso necesaria para poder proceder al suministro del producto.
Este producto ha sido bloqueado por su organización. Por favor, pónganse en contacto con su departamento de compras para obtener más información.
El producto original ya no está disponible. Su sustituto se muestra a continuación.
El producto(s) marcados con este símbolo están descatalogados - se venden hasta acabar stock. Pueden encontrar alternativas buscando con el número de catálogo VWR listado arriba. Si necesita más ayuda, por favor llame al Servicio de atención al cliente de VWR al 902.222.897.
1 - 16 of 27
no targeter for Bottom