Safety spectacles, Fuel X2
Proveedor: 3M
MMMAFUELX20EA 18 EUR
MMMAFUELX20
Safety spectacles, Fuel X2
Safety glasses 'Fuel' combine comfort and style efficiently.
  • Adjustable rubber straps
  • Soft, individually adjustable nose bridge
Weight: 29 g
Certificaciones: EN 166 5-3.1 1 FT; EN 170; EN 172
Información de suministro: With a microfibre pouch to store and clean the glasses.