DNA transfection reagent, jetPEI®-HUVEC
Proveedor: Polyplus-transfection
108-05NEA 439 EUR
PPLU108-05N
DNA transfection reagent, jetPEI®-HUVEC
jetPEI®-HUVEC is a powerful transfection reagent optimised for the transfection of primary human endothelial cells, such as HUVEC (Human umbilical vein endothelial cells).
  • Efficient transfection of endothelial cells (up to 50% in HUVEC)
  • As efficient as electroporation
  • Easy-to-use protocol
  • Good cell viability
  • Compatible with serum and antibiotics
Transfection efficiencies up to 50% have been reached with this reagent. jetPEI®-HUVEC is also recommended for the transfection of vascular endothelial cells of various origins and appears to be well suited for such fragile primary cells.
Certificaciones: Polyplus-transfection® has been an ISO 9001 accredited company since 2002. Since then this certification has been successfully renewed every three years.
Every batch of jetPEI®-HUVEC is tested in a transfection assay. Typically, transfection of a firefly luciferase gene under the control of the CMV promoter gives 109 RLU (relative light unit)/mg of protein. The value for each batch is indicated on the Certificate of Analysis.
Información de suministro: 150 mM NaCl solution is included. 0,5 ml of jetPEI®-HUVEC transfection reagent is sufficient to perform up to 125 transfections in 24-well plates.