Rotary evaporator, Rotavapor® R-210
Proveedor: Büchi
Rotavapor®
531-1900EA 0 EUR
531-1900 531-1901 531-1902 531-0089 531-0240 531-0241 531-0103 531-0268 531-1896 531-1897 531-0269 531-0104 531-0247 531-0101 531-0129 531-1898 531-0249 531-1899 531-1916 531-0924 531-0275 531-0254 531-0251 531-0097 531-0273 531-0274 531-0252 531-0255 531-0850 531-0256 531-0092 531-0271 531-0250 531-0272 531-0270
Rotary evaporator, Rotavapor® R-210
Evaporadores Rotavapores
Rotavapor® R-210 rotary evaporator for rapid and complex distillation of all types of solvents. Works with evaporator flasks of 50 to 4000 ml. The R-210 is equipped with an electric lifting and lowering device and automatic lifting of the evaporator flask in the event of a power failure.
  • Electronically adjustable speed
  • Heating bath designed to heat water or oil within a temperature range of +20 to +180 °C
  • Heating bath comes with a digital temperature display fitted as standard
Rotavapor® R-210 Advanced
The R-210 Advanced comes with a V-850 vacuum controller that is used to control a V700 or V710 pump and which has a timer function and program memory. Rotating and lifting can be controlled by the controller. The integrated library containing 43 solvents, which can be updated via USB, facilitates fast and efficient evaporation.
Rotavapor® R-210 Professional**
The R-210 Professional includes the V-855 vacuum controller. You can choose different settings, such as continuous vacuum, pressure gradients, timer functions or complete automatic distillation. Integrated EasyVac and LabVac functions. Rotating and lifting can be controlled. The integrated library containing 43 solvents, which can be updated via USB, facilitates fast and efficient evaporation. Please contact your local Avantor sales office for details.
Glassware
Glassware A, diagonal condenser: For standard distillations, can be used in a confined space, evaporator flask feed.
Glassware V, vertical condenser: For standard distillations, evaporator flask feed, connection for vapour temperature sensor, automatic distillation possible with the V-855 vacuum controller.
Glassware C, dry ice condenser: For distilling solvents with low boiling points, evaporator flask feed, no cooling liquid necessary.
Glassware S, vertical condenser with shut-off valve: For standard distillations, evaporator flask feed, connection for vapour temperature sensor, automatic distillation possible with V-855 vacuum controller.
Glassware E, descending condenser with expansion vessel: Ideal for distilling substances that have a tendency to foam and that take a long time to reach boiling point, evaporator flask feed, connection for vapour temperature sensor.
Glassware BY, intensive condenser: Vertical condenser with double mantle, additional joint on the top of the condenser to allow expansion, evaporator flask feed, connection for vapour temperature sensor.
Información para pedidos: Supplied complete and ready to use with 1000 ml receiving flask, 1000 ml evaporating flask and heating bath.
Please note: A vacuum source, vacuum check valve and cooling system are required but not supplied. Please contact your local Avantor sales office for advice and details.
Precaución: * With transparent plastic coating
** The valve unit (181-0140) or the Woulff bottle (181-0141) (to be used with a V-700 or V-710 vacuum pump) are available as optional extras.