1-Metil-3-nitroguanidina
Proveedor: EHRENSTORFER GMBH
EHERC15106500EA
116
EUR
EHERC15106500
1-Metil-3-nitroguanidina
1-Metil-3-nitroguanidina
Formula:
C₂H₅N₅O₃ MW: 147,09 g/mol |
Número MDL:
MFCD00024167 Núm. CAS: 4245-76-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...