Vacuum pump, Stuart®, RE3022C
Proveedor: Cole Parmer
STUARE3022CEA 4040 EUR
STUARE3022C
Vacuum pump, Stuart®, RE3022C
Bombas Bombas de rotavapores de vacío
The RE3022C model is a powerful, compact vacuum pump, constructed with solid PTFE heads, PTFE diaphragms and Kalrez® valves. The pump is designed to withstand the challenging role of a chemical laboratory pump.
  • Quiet operation with no vibration
  • Vacuum regulator for easy adjustment
The pump is useful for any applications requiring vacuum generation and is ideal to be used with rotary evaporator. By utilising a pump to place the rotary evaporator under vacuum, distillation efficiency is greatly improved. By reducing the pressure within the glassware of the rotary evaporator solvents can be evaporated off at lower temperatures ensuring the bath can be kept at a safer temperature.
Información de suministro: Supplied with vacuum regulator and catch pots to protect the pump from liquid ingress, as standard.