Digesters DK series
Proveedor: VELP Scientifica
VELPF30100360EA 2480 EUR
VELPF30100360 VELPF30100350
Digesters DK series
A range of digesters for use in laboratories performing analysis of diverse applications in food and feed, beverage (nitrogen, protein, Total Kjeldahl Nitrogen), environmental (COD, Total Kjeldahl Nitrogen), chemical and pharmaceutical (organic nitrogen) industries, according to a variety of standards (such as AOAC, ISO, EPA, DIN etc).
  • Simple to use with just four keys to set the required operations
  • Programmable temperature ramps
  • Rapid heating rate 20 to 420 °C in 30 minutes
  • Electronic microprocessor checks and automatically calibrates the aluminium heating block temperature
  • Choice of six different languages