Automated microtomes, RM 2255
Proveedor: Leica
720-1882EA 0 EUR
720-1882 720-2267
Automated microtomes, RM 2255
Microtomas Micrótomos
The RM 2255 perfectly harmonises practical features with an ergonomic design and user safety. Its two-in-one design concept allows motorised as well as manual sectioning with two forward and backward coarse feed speeds. Speed control through the cutting window for enhanced efficiency and automatic, variable specimen retraction, depending on sectioning speed.
  • Powerful sectioning motor rugged for sectioning most hard specimens
  • Special electronics system allows flexible operation while providing precision control over the specimen advance and cutting speeds
  • Patented gravitational force compensation system makes rotating the hand wheel extremely smooth and light, which is especially beneficial during manual sectioning applications
  • One-hand operated universal cassette clamp is an easy way to clamp and remove specimen
Rotary microtome for high-performance motorised and manual sectioning.
Información para pedidos: The set includes precision orientation, quick clamping system for specimen holders, universal cassette clamp, knife holder base - without lateral displacement feature, knife holder E for low-profile disposable blades and storage tray.