Microtome, RM 2125 RTS
Proveedor: Leica
720-2264EA 11510 EUR
720-2264 720-1880 LEIH14045746960
Microtome, RM 2125 RTS
Microtomas Micrótomos
The RM2125 RTS has the essential features for safer sectioning and optimised workflows. A quick-exchange specimen clamp and an 8° orientation system for the object head allow rapid set up for any block. Specimen retraction: ON/OFF and coarse feed wheel turn direction is user selectable.
  • Accurate feed mechanism for precise sections
  • PowerBase for stable operation and vibration-free sectioning
  • Secure advanced knife holder and specimen clamp securely align cutting edge to block
  • Productivity – Two trim steps (50 μm coarse, 10 μm fine) mean fast trimming and store essential tools and accessories on top of the microtome so they are easy to access