Compact automatic burettes, Schellbach stripe, BLAUBRAND®
Proveedor: Brand
BRND23919EA 156 EUR
BRND23919 BRND23921 BRND23920
Compact automatic burettes, Schellbach stripe, BLAUBRAND®
Borosilicate glass 3.3, class AS, DE-M marking.
  • Modular concept - all components, including PTFE stopcock, are replaceable
  • Calibrated to deliver 'TD, Ex'
  • Automatic zeroing and Schellbach stripe
Certificaciones: DIN EN ISO 385
Información para pedidos: Supplied with a batch certificate. Burettes are also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate. Please order burette clamps,supports and rubber bellows separately.
Información de suministro: Burette supplied with 2000 ml reservoir bottle, pumphead and PVC filling tube.