¿Quiere conectarse de nuevo?
Dispersers, LABOR-PILOT 2000/4
Proveedor: IKA
DISPAX-REACTOR®
The LABOR-PILOT is a compact and multi-purpose inline laboratory dispersing machine. It is designed for solid-liquid mixing, homogenising, emulsifying, suspending and wet milling. The LABOR-PILOT 2000/4 is suitable for wide range of applications, particularly in the chemical, cosmetic, pharmaceutical, plastics, colours and food industries. It is uniquely designed with advanced technology and is available with six interchangeable modules.
  • Flexibility and ease of use: One machine suits for many applications and variety of processes
  • Together with inline machines of 2000 series enables seamless transition of processes from laboratory to production while retaining the product quality
  • Easy and quick exchange of the modules as well as conversion to a complete working plant
  • CIP and SIP capable
  • Integral electronic control via LABOR-PILOT-CONTROLLER
It can be customized to fit particular procedural demands. Important process parameters such as circumferential speed, tools and mixing chamber geometry are structurally identical to those of the IKA 2000 inline production machines: UTL, DR, DRS, MHD, MK and MKO. Therefore, the LABOR-PILOT ensures a reliable scale-up by offering the possibility to work with the same method from formulation development to mass production. The laboratory dispersing machine LABOR-PILOT 2000/4 can be upgraded to a high-speed dispersing machine with the LABOR-PILOT-CONTROLLER.
The LABOR-PILOT 2000/4 differs from the alternative PROCESS-PILOT 2000/4 machine due to the applied seal technology and two additional process modules for powder entry in recirculation mode. For working under pressure/vacuum and with elevated temperatures or abrasive materials, we recommend the machine PROCESS-PILOT. The basic inline machine is equipped with the single-stage dispersing module ULTRA-TURRAX® UTL. It is best suited for batch applications and can be adapted for the manufacturing of dispersions from coarse to fine. Compared to a conventional stirrer, the mixing time is reduced by up to 50%. Depending on the application, the basic unit can be modified quickly with different working modules.
The DISPAX-REACTOR® DR module converts the LABOR-PILOT into a multi-stage high capacity dispersing machine for fine particle size reduction. With just one pass, the module DR does typically achieve fine dispersions with an extremely narrow particle size distribution. It also guarantees an optimum homogeneity, which is an important pre-condition for the long-time stability of emulsions or suspensions.
The MHD module is designed for a fully continuous incorporation of powders into liquids. By use of this module, the mixing relation of solid and liquid should be volumetrically or gravimetrically controlled. Depending on the product characteristics, dispersions with viscosities up to 50 Pas or solid contents up to 80% can be achieved just after one pass.
The Colloid Mill MK and Cone Mill MKO modules are used for wet or fine milling of hard and granular raw materials or for the production of finest emulsions as well as for improving of quality of finished dispersions. The MK module consists of two cones with differently directed flow channels on their surfaces. The cones can be shifted against each other to a minimum distance, thus varying friction as well as flow capacity. By adjusting the milling gap, the achievable particle size can be controlled. The MKO module works on the same principle as the MK module except the outlet area of its cones is coated with abrasion-resistant tungsten carbide. This results in an enlarged friction surface that enables to produce even finer suspensions at reduced flow.
Utilice su UserId y contraseña para ver su precio y disponibilidad.
  Recordar mis datos en este dispositivo
PEDIDO
Abrir todos / Contraer todo
Descripción Env Núm. Catálogo VWR Cantidad
Dispersers, LABOR-PILOT with clamp connection 1 IKAAU103694
LABOR-PILOT Controller
Descripción Env Núm. Catálogo VWR Cantidad
Controlador 1,5 kW, LABOR-PILOT 1 IKAAT054916
Modules
Descripción Env Núm. Catálogo VWR Cantidad
Módulo DISPAX-REACTOR® para LABOR-PILOT 1 IKAAT055013
Módulo MHD para LABOR-PILOT 1 IKAAT055142
Módulo MKO para LABOR-PILOT 1 IKAAT061069
Módulo MK para LABOR-PILOT 1 IKAAT054917
Generate Barcodes Barcode Label Format: 
Generar página de catálogo en PDF
ESPECIFICACIONES
Modelo LABOR-PILOT 2000/4
Volumen de H2O (ml) Flow capacity: 300 - 700 l/h
Intervalo de velocidad (min-1) 3160 - 13750 (with PILOT controller)
Potencia del motor (W) 1500
Ancho × Fondo × Alto (mm) 450×250×350
Peso (kg) 36
Enter Details
Select the article for which you are looking for Certificate
Indicar núm. de lote (campo obligatorio). Introduzca 'x' si lo desconoce para productos VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
O
Revise y compare las alternativas
Disponemos de alternativas con las que puede ahorrar hasta  por producto unitario.  Para comparar detalles de los productos, seleccione hasta 3 alternativas y haga clic en comparar seleccionados. Para añadir productos a su cesta, introduzca la cantidad y haga clic en Añadir a la cesta.
Producto Original:
Descripción Numero del catalogo Disponibilidad Unidad Su precio Precio Por Uds.