2-Metil-4(5)-nitroimidazol
Proveedor: LGC Standards PROMOCHEM
Sinónimos:
2-Metil-5-nitroimidazol
, 2-Metil-4-nitroimidazol
, 2-Metil-4-nitro-1H-imidazol
PROOBP244EA
247
EUR
PROOBP244
2-Metil-4(5)-nitroimidazol
2-Metil-4(5)-nitroimidazol
Formula:
C₄H₅N₃O MW: 127,10 g/mol |
Número MDL:
MFCD00005191 Núm. CAS: 696-23-1 EINECS: 211-790-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...