Esta buscando: Biología humana
129  results were found
SearchResultCount:"129"
Sort Results
Vista lista Vista Extendida (Nueva)
Valore los resultados de su búsqueda
Proveedor: Binhold, Paul Lehrmittel
Descripción: Teach proper cleaning techniques using this giant toothbrush.
Numero del catalogo: (763-0124)
Proveedor: Binhold, Paul Lehrmittel
Descripción: Three models on a common plate. Kidney in cross section enlarged 3×. Nephron enlarged 120× and kidney capsule enlarged 700×. Dimensions: 29×52×9 cm.
UOM: 1 * 1 UN
Market Source Item This is a MarketSource item. Additional charges may apply
Numero del catalogo: (BINH1000194)
Proveedor: Binhold, Paul Lehrmittel
Descripción: This human mini torso is approximately half life-size, supplied with the 3B Torso Guide. Even small hands can quickly disassemble and assemble this to...
UOM: 1 * 1 UN
Market Source Item This is a MarketSource item. Additional charges may apply
Proveedor: Binhold, Paul Lehrmittel
Descripción: All important structures of the human kidney for student and patient education are shown.
Proveedor: Binhold, Paul Lehrmittel
Descripción: This full-size manikin allows the training of BLS rescue techniques and CPR. The resiliency and weight of the body is very realistic.
Proveedor: Binhold, Paul Lehrmittel
Descripción: Modelo molecular para fines educativos, Muscle Layer
Proveedor: Binhold, Paul Lehrmittel
Descripción: See the complete stages of development. Study the normal position of child before birth with this model plus the human reproductive and urinary system...
Proveedor: Binhold, Paul Lehrmittel
Descripción: Life-size relief model with the internal organs of the head. With hanger.
Numero del catalogo: (BINH1000112)
Proveedor: Binhold, Paul Lehrmittel
Descripción: This model is a realistic replica of the human clavicle.
UOM: 1 * 1 UN
Market Source Item This is a MarketSource item. Additional charges may apply
Proveedor: Binhold, Paul Lehrmittel
Descripción: These models allow trainees to practice both basic and medical infant care.
Proveedor: Binhold, Paul Lehrmittel
Descripción: This anatomy model shows one half of the female and male genital organs with bladder and removable rectum.
Proveedor: Binhold, Paul Lehrmittel
Descripción: Simulators are ideal for education.
Proveedor: Binhold, Paul Lehrmittel
Descripción: Reduced to the natural size, these human models keep all of the functionality.
Numero del catalogo: (BINH1000025)
Proveedor: Binhold, Paul Lehrmittel
Descripción: The replica of the human dis-articulated skeleton includes one hand and foot on wire, and one hand and foot loosely articulated.
UOM: 1 * 1 UN
Market Source Item This is a MarketSource item. Additional charges may apply
Numero del catalogo: (763-0134)
Proveedor: Binhold, Paul Lehrmittel
Descripción: Muscles, ligaments, blood vessels and nerves of the legs and feet.
UOM: 1 * 1 UN
Market Source Item This is a MarketSource item. Additional charges may apply
Numero del catalogo: (BINH1000211)
Proveedor: Binhold, Paul Lehrmittel
Descripción: This female muscular figure includes the following removable parts:
UOM: 1 * 1 UN
Market Source Item This is a MarketSource item. Additional charges may apply
Consulta de precio
El stock para este material es limitada pero puede estar disponible en un almacén cerca de usted. Por favor, asegúrese de que ha iniciado sesión en la web para que el stock disponible se puede mostrar. Si el call sigue apareciendo y usted necesita ayuda, por favor llámenos al 902 222 897 o por email en webshop.es@avantorsciences.com.
El stock para este material es limitada pero puede estar disponible en un almacén cerca de usted. Por favor, asegúrese de que ha iniciado sesión en la web para que el stock disponible se puede mostrar. Si el call sigue apareciendo y usted necesita ayuda, por favor llámenos al 902 222 897 o por email en webshop.es@avantorsciences.com.
Este producto se trata de un artículo regulado sometido a normativa que restringe su venta. Si procede, nos pondremos en contacto con usted para solicitarle la licencia o declaración de uso necesaria para poder proceder al suministro del producto.
Este producto se trata de un artículo regulado sometido a normativa que restringe su venta.
Si procede, nos pondremos en contacto con usted para solicitarle la licencia o declaración de uso necesaria para poder proceder al suministro del producto.
Este producto ha sido bloqueado por su organización. Por favor, pónganse en contacto con su departamento de compras para obtener más información.
El producto original ya no está disponible. Su sustituto se muestra a continuación.
El producto(s) marcados con este símbolo están descatalogados - se venden hasta acabar stock. Pueden encontrar alternativas buscando con el número de catálogo VWR listado arriba. Si necesita más ayuda, por favor llame al Servicio de atención al cliente de VWR al 902.222.897.
1 - 16 of 129
no targeter for Bottom