Esta buscando: Bombas
517  results were found
SearchResultCount:"517"
Sort Results
Vista lista Vista Extendida (Nueva)
Valore los resultados de su búsqueda
Proveedor: Merck
Descripción: Merck offers a broad variety of withdrawal systems that are tailor-made for barrels and drums. They are designed to ensure safe, easy and efficient so...
Proveedor: Avantor Fluid Handling
Descripción: Designed with maintenance-free high-accuracy motor and a graphical interface—makes setup and operation easier than ever!
CE Compliant
Proveedor: Avantor Fluid Handling
Descripción: Independently control flow and direction of each channel via keypad or your PC.
Proveedor: Avantor Fluid Handling
Descripción: Versatile multichannel pumps for accurate low-flow fluid delivery – with an intuitive touch-screen interface.
MasterflexLive®
Proveedor: Avantor Fluid Handling
Descripción: Networking drives offer either native EtherNet/IP™ or Profibus communication—no additional hardware or firmware required.
Proveedor: Avantor Fluid Handling
Descripción: Compact, cost effective pump systems for general fluid transfer applications.
Proveedor: Avantor Fluid Handling
Descripción: Extend the life of your gear pump head with these service kits.
Proveedor: Avantor Fluid Handling
Descripción: Achieve high pressure, accuracy, and repeatability—with an intuitive touch-screen interface.
Numero del catalogo: (99900.001)
Proveedor: VWR Chemicals
Descripción: Specially developed solvent withdrawal system with manual pressure build-up for VWR® returnable 10 and 30 L stainless steel barrels.
UOM: 1 * 1 UN
Proveedor: Avantor Fluid Handling
Descripción: Make the most of your Masterflex drives replacement parts and other accessories.
Proveedor: Avantor Fluid Handling
Descripción: Accommodate up to two syringes in a cost-effective touch-screen syringe pump.
CE Compliant CSA Certified UL Listed
Proveedor: Avantor Fluid Handling
Descripción: Easy operation and easy
on the budget.
Proveedor: Avantor Fluid Handling
Descripción: Smooth, precise flow and fewer tubing changes.
CE Compliant
Proveedor: Avantor Fluid Handling
Descripción: Every drop counts—integrate into automated filling systems for high accuracy dosing and dispensing.
Proveedor: Avantor Fluid Handling
Descripción: Control flow rate via the user-friendly touch-screen display.
UL Listed
Proveedor: Avantor Fluid Handling
Descripción: Ideal for the transfer and dispensing of live cell cultures, media, and other shear-sensitive fluids in process and washdown environments.
Consulta de precio
El stock para este material es limitada pero puede estar disponible en un almacén cerca de usted. Por favor, asegúrese de que ha iniciado sesión en la web para que el stock disponible se puede mostrar. Si el call sigue apareciendo y usted necesita ayuda, por favor llámenos al 902 222 897 o por email en webshop.es@avantorsciences.com.
El stock para este material es limitada pero puede estar disponible en un almacén cerca de usted. Por favor, asegúrese de que ha iniciado sesión en la web para que el stock disponible se puede mostrar. Si el call sigue apareciendo y usted necesita ayuda, por favor llámenos al 902 222 897 o por email en webshop.es@avantorsciences.com.
Este producto se trata de un artículo regulado sometido a normativa que restringe su venta. Si procede, nos pondremos en contacto con usted para solicitarle la licencia o declaración de uso necesaria para poder proceder al suministro del producto.
Este producto se trata de un artículo regulado sometido a normativa que restringe su venta.
Si procede, nos pondremos en contacto con usted para solicitarle la licencia o declaración de uso necesaria para poder proceder al suministro del producto.
Este producto ha sido bloqueado por su organización. Por favor, pónganse en contacto con su departamento de compras para obtener más información.
El producto original ya no está disponible. Su sustituto se muestra a continuación.
El producto(s) marcados con este símbolo están descatalogados - se venden hasta acabar stock. Pueden encontrar alternativas buscando con el número de catálogo VWR listado arriba. Si necesita más ayuda, por favor llame al Servicio de atención al cliente de VWR al 902.222.897.
1 - 16 of 517
no targeter for Bottom