Safety covers for block heaters, stuart®
Proveedor: Cole Parmer
Stuart®
460-0001EA 224 EUR
460-0001 460-0000
Safety covers for block heaters, stuart®
Cubiertas
Tough Perspex® covers for Stuart block heaters.
  • Easy to fit, does not require tools
  • Protect user for possible spitting tubes
  • Improve temperature stability by preventing drafts