Esta buscando: Sistemas de flujo tangencial
19  results were found
SearchResultCount:"19"
Sort Results
Vista lista Vista Extendida (Nueva)
Valore los resultados de su búsqueda
Proveedor: Cytiva (Formerly Pall Lab)
Descripción: Sistema de flujo tangencial, Sistema Centramate™, incluye soporte de casete para Centramate™, surtido de adaptadores, llave dinamométrica y enchufe
Proveedor: SPECTRUM LABORATORIES
Descripción: Hollow fibre filter modules are particularly useful in tangential flow filtration because processes can easily be scaled-up from small volume applicat...
Proveedor: Cytiva (Formerly Pall Lab)
Descripción: These systems are designed for use in diafiltration procedures which are faster and more efficient than dialysis for buffer exchange and desalting, an...
Numero del catalogo: (516-1085)
Proveedor: Cytiva (Formerly Pall Lab)
Descripción: Reusable TFF devices and system for bioprocessing applications that accelerates and simplifies processing of up to 1 L.
UOM: 1 * 1 UN
Numero del catalogo: (516-8200)
Proveedor: Cytiva (Formerly Pall Lab)
Descripción: Ultralab™ systems, composed of Ultrareservoir™ containers and Ultrapump™ II pumps, are designed as a rapid, efficient replacement of dialysis for buff...
UOM: 1 * 1 UN
Proveedor: SPECTRUM LABORATORIES
Descripción: Designed for gentle tangential flow filtration of small volumes ranging from 1 to 100 ml. MicroKros® modules are typically operated by a peristaltic p...
Proveedor: SPECTRUM LABORATORIES
Descripción: Hollow fibre filter modules are particularly useful in tangential flow filtration because processes can easily be scaled-up from small volume applicat...
Proveedor: SPECTRUM LABORATORIES
Descripción: Hollow fibre filter modules are particularly useful in tangential flow filtration because processes can easily be scaled-up from small volume applicat...
Proveedor: SPECTRUM LABORATORIES
Descripción: Hollow fibre filter modules are particularly useful in tangential flow filtration because processes can easily be scaled-up from small volume applicat...
Proveedor: SPECTRUM LABORATORIES
Descripción: Hollow fibre filter modules are particularly useful in tangential flow filtration because processes can easily be scaled-up from small volume applicat...
Proveedor: SPECTRUM LABORATORIES
Descripción: Hollow fibre filter modules are particularly useful in tangential flow filtration because processes can easily be scaled-up from small volume applicat...
Proveedor: SPECTRUM LABORATORIES
Descripción: Sistema de flujo tangencial, Fibre filter, mPES, for 10 - 100 L volumes
Proveedor: SPECTRUM LABORATORIES
Descripción: Hollow fibre filter modules are particularly useful in tangential flow filtration because processes can easily be scaled-up from small volume applicat...
New Product
Proveedor: SPECTRUM LABORATORIES
Descripción: Hollow fibre filter modules are particularly useful in tangential flow filtration because processes can easily be scaled-up from small volume applicat...
Proveedor: SPECTRUM LABORATORIES
Descripción: Hollow fibre filter modules are particularly useful in tangential flow filtration because processes can easily be scaled-up from small volume applicat...
Proveedor: Sartorius
Descripción: Vivaflow® 200 is a ready to use and reusable laboratory crossflow cassette for concentrating up to 5 L samples.
Consulta de precio
El stock para este material es limitada pero puede estar disponible en un almacén cerca de usted. Por favor, asegúrese de que ha iniciado sesión en la web para que el stock disponible se puede mostrar. Si el call sigue apareciendo y usted necesita ayuda, por favor llámenos al 902 222 897 o por email en webshop.es@avantorsciences.com.
El stock para este material es limitada pero puede estar disponible en un almacén cerca de usted. Por favor, asegúrese de que ha iniciado sesión en la web para que el stock disponible se puede mostrar. Si el call sigue apareciendo y usted necesita ayuda, por favor llámenos al 902 222 897 o por email en webshop.es@avantorsciences.com.
Este producto se trata de un artículo regulado sometido a normativa que restringe su venta. Si procede, nos pondremos en contacto con usted para solicitarle la licencia o declaración de uso necesaria para poder proceder al suministro del producto.
Este producto se trata de un artículo regulado sometido a normativa que restringe su venta.
Si procede, nos pondremos en contacto con usted para solicitarle la licencia o declaración de uso necesaria para poder proceder al suministro del producto.
Este producto ha sido bloqueado por su organización. Por favor, pónganse en contacto con su departamento de compras para obtener más información.
El producto original ya no está disponible. Su sustituto se muestra a continuación.
El producto(s) marcados con este símbolo están descatalogados - se venden hasta acabar stock. Pueden encontrar alternativas buscando con el número de catálogo VWR listado arriba. Si necesita más ayuda, por favor llame al Servicio de atención al cliente de VWR al 902.222.897.
1 - 16 of 19
no targeter for Bottom