4-Benzyl-2-(hydroxymethyl)piperazine 95%

Proveedor: BLD PHARMATECH GMBH

BLDPBD99257-1GEA 148 EUR
BLDPBD99257-1G BLDPBD99257-250MG BLDPBD99257-5G
4-Benzyl-2-(hydroxymethyl)piperazine 95%
4-Benzyl-2-(hydroxymethyl)piperazine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₁₂H₁₈N₂O
MW: 206,284 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD03001699
Núm. CAS: 85817-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR