4-Methoxy-3-nitrobenzonitrile 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD98437-25GEA 293 EUR
BLDPBD98437-25G BLDPBD98437-5G BLDPBD98437-0.025G BLDPBD98437-250MG BLDPBD98437-1G
4-Methoxy-3-nitrobenzonitrile 98%
4-Methoxy-3-nitrobenzonitrile

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₆N₂O₃
MW: 178,1448 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00221439
Núm. CAS: 33224-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR