2-Bromo-6-fluorophenol 98%

BLDPBD9813-100GEA 251 EUR
BLDPBD9813-100G BLDPBD9813-1G BLDPBD9813-25G BLDPBD9813-10G BLDPBD9813-5G
2-Bromo-6-fluorophenol 98%
2-Bromo-6-fluorophenol

Storage condition: Inert atmosphere, 2-8°C

Formula: C₆H₄BrFO
MW: 191,0000 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD03788533
Núm. CAS: 2040-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR