3-Amino-4-iodo-1H-indazole 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD98049-5GEA 112 EUR
BLDPBD98049-5G BLDPBD98049-1G
3-Amino-4-iodo-1H-indazole 97%
3-Amino-4-iodo-1H-indazole

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₇H₆IN₃
MW: 259,0471 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD03426697
Núm. CAS: 599191-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR