2-Chloro-4-fluoro-5-nitrophenol 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD9772-5GEA 35 EUR
BLDPBD9772-5G BLDPBD9772-25G BLDPBD9772-1G
2-Chloro-4-fluoro-5-nitrophenol 97%
2-Chloro-4-fluoro-5-nitrophenol

Storage condition: Inert atmosphere, Room Temperature

Formula: C₆H₃ClFNO₃
MW: 191,5400 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD02670258
Núm. CAS: 84478-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR