4'-Chloro-2'-fluoro-5'-methylacetophenone 95%

BLDPBD95709-25GEA 247 EUR
BLDPBD95709-25G BLDPBD95709-1G BLDPBD95709-5G BLDPBD95709-250MG
4'-Chloro-2'-fluoro-5'-methylacetophenone 95%
4'-Chloro-2'-fluoro-5'-methylacetophenone

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₈ClFO
MW: 186,6106 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00236601
Núm. CAS: 177211-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR