4-Chloro-2,6-difluorobenzaldehyde 97%
|
|
BLDPBD95674-100MGEA
14
EUR
BLDPBD95674-100MG
BLDPBD95674-250MG
BLDPBD95674-5G
BLDPBD95674-0.025G
BLDPBD95674-25G
BLDPBD95674-1G
4-Chloro-2,6-difluorobenzaldehyde 97%
4-Chloro-2,6-difluorobenzaldehyde
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₇H₃ClF₂O MW: 176,55 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD03094460 Núm. CAS: 252004-45-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...