4-Bromo-N-phenylaniline 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD95236-5GEA 22 EUR
BLDPBD95236-5G BLDPBD95236-25G BLDPBD95236-1G
4-Bromo-N-phenylaniline 98%
4-Bromo-N-phenylaniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₂H₁₀BrN
MW: 248,1200 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD07779504
Núm. CAS: 54446-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR