4-Bromobenzylamine hydrochloride 95%

BLDPBD95211-5GEA 16 EUR
BLDPBD95211-5G BLDPBD95211-100G BLDPBD95211-10G BLDPBD95211-1G BLDPBD95211-0.025G BLDPBD95211-25G
4-Bromobenzylamine hydrochloride 95%
4-Bromobenzylamine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₉BrClN
MW: 222,52 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00012860
Núm. CAS: 26177-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR