(±)-2-Piperazinecarboxylic acid dihydrochloride 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD9476-25GEA 49 EUR
BLDPBD9476-25G BLDPBD9476-10G BLDPBD9476-5G BLDPBD9476-1G BLDPBD9476-100G
(±)-2-Piperazinecarboxylic acid dihydrochloride 98%
(±)-2-Piperazinecarboxylic acid dihydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₅H₁₂Cl₂N₂O₂
MW: 203,07 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00044803
Núm. CAS: 3022-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR