4,4'-Biphenyldiboronic acid bis(neopentyl glycol)ester 98%

BLDPBD91993-5GEA 170 EUR
BLDPBD91993-0.025G BLDPBD91993-5G BLDPBD91993-1G BLDPBD91993-25G BLDPBD91993-250MG
4,4'-Biphenyldiboronic acid bis(neopentyl glycol)ester 98%
4,4'-Biphenyldiboronic acid bis(neopentyl glycol)ester

Storage condition: Sealed in dry, Room Temperature

Formula: C₂₂H₂₈B₂O₄
MW: 378,0773 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD01318103
Núm. CAS: 5487-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR