4-(tert-Butylcarbamoyl)phenylboronic acid 98%
|
|
BLDPBD91638-1GEA
35
EUR
BLDPBD91638-1G
BLDPBD91638-25G
BLDPBD91638-250MG
BLDPBD91638-0.025G
BLDPBD91638-5G
4-(tert-Butylcarbamoyl)phenylboronic acid 98%
4-(tert-Butylcarbamoyl)phenylboronic acid
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₁₁H₁₆BNO₃ MW: 221,06 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD03788413 Núm. CAS: 850568-14-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...