(R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD9060-250MGEA 6 EUR
BLDPBD9060-250MG BLDPBD9060-10G BLDPBD9060-5G BLDPBD9060-25G BLDPBD9060-500G BLDPBD9060-100G BLDPBD9060-1G
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate 97%
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₂₀H₁₃O₄P
MW: 348,289 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00010045
Núm. CAS: 39648-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR