4-(3-Chloroprop-1-yl)morpholine hydrochloride 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD90237-100MGEA 6 EUR
BLDPBD90237-100MG BLDPBD90237-250MG BLDPBD90237-1G
4-(3-Chloroprop-1-yl)morpholine hydrochloride 98%
4-(3-Chloroprop-1-yl)morpholine hydrochloride

Storage condition: Sealed in dry, 2-8°C

Formula: C₇H₁₄ClNO·ClH
MW: 200,106 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00035333
Núm. CAS: 57616-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR