4-Bromo-4'-pentylbiphenyl 98%

BLDPBD8878-250MGEA 7 EUR
BLDPBD8878-250MG BLDPBD8878-1G BLDPBD8878-10G BLDPBD8878-25G BLDPBD8878-100G BLDPBD8878-5G
4-Bromo-4'-pentylbiphenyl 98%
4-Bromo-4'-pentylbiphenyl

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₇H₁₉Br
MW: 303,2400 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00060106
Núm. CAS: 63619-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR