3-Butoxypropionitrile 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD88424-25GEA 19 EUR
BLDPBD88424-25G BLDPBD88424-100G BLDPBD88424-5G
3-Butoxypropionitrile 98%
3-Butoxypropionitrile

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₁₃NO
MW: 127,19 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00045643
Núm. CAS: 6959-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR