5-Nitro-m-cresol 98%

BLDPBD88108-250MGEA 74 EUR
BLDPBD88108-250MG BLDPBD88108-100MG
5-Nitro-m-cresol 98%
5-Nitro-m-cresol

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₇NO₃
MW: 153,1354 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD09038591
Núm. CAS: 127818-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR