3-Methoxypyridazine 97%
Proveedor: BLD PHARMATECH GMBH
|
|
BLDPBD87838-5GEA
108
EUR
BLDPBD87838-0.025G
BLDPBD87838-5G
BLDPBD87838-1G
BLDPBD87838-25G
BLDPBD87838-250MG
BLDPBD87838-100MG
3-Methoxypyridazine 97%
3-Methoxypyridazine
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₅H₆N₂O MW: 110,1139 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00234098 Núm. CAS: 19064-65-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...