5-Methoxy-3-pyridineboronic acid pinacol ester 97%

BLDPBD87736-1GEA 17 EUR
BLDPBD87736-1G BLDPBD87736-250MG BLDPBD87736-10G BLDPBD87736-5G
5-Methoxy-3-pyridineboronic acid pinacol ester 97%
5-Methoxy-3-pyridineboronic acid pinacol ester
Formula: C₁₂H₁₈BNO₃
MW: 235,0872 g/mol
Número MDL: MFCD03412068
Núm. CAS: 445264-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR