3-Ethoxypropionitrile 99%

Proveedor: BLD PHARMATECH GMBH

BLDPBD86579-100GEA 17 EUR
BLDPBD86579-100G BLDPBD86579-1000G BLDPBD86579-500G BLDPBD86579-25G
3-Ethoxypropionitrile 99%
3-Ethoxypropionitrile

Storage condition: Sealed in dry, Room Temperature

Formula: C₅H₉NO
MW: 99,1300 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00001959
Núm. CAS: 2141-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR