3-Chloro-6-iodopyridazine 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD85994-1GEA 13 EUR
BLDPBD85994-1G BLDPBD85994-250MG BLDPBD85994-25G BLDPBD85994-5G BLDPBD85994-10G
3-Chloro-6-iodopyridazine 97%
3-Chloro-6-iodopyridazine
Formula: C₄H₂ClIN₂
MW: 240,43 g/mol
Número MDL: MFCD08275187
Núm. CAS: 135034-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR