N-Fmoc-glycine 98%
|
|
BLDPBD8594-10GEA
9
EUR
BLDPBD8594-10G
BLDPBD8594-0.025G
BLDPBD8594-500G
BLDPBD8594-1G
BLDPBD8594-5G
BLDPBD8594-100G
BLDPBD8594-25G
N-Fmoc-glycine 98%
N-Fmoc-glycine
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₁₇H₁₅NO₄ MW: 297,3 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00037140 Núm. CAS: 29022-11-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...