3-Bromo-5-cyano-1H-indole, N-BOC protected 98%

BLDPBD85174-5GEA 183 EUR
BLDPBD85174-5G BLDPBD85174-250MG BLDPBD85174-1G
3-Bromo-5-cyano-1H-indole, N-BOC protected 98%
3-Bromo-5-cyano-1H-indole, N-BOC protected

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₄H₁₃BrN₂O₂
MW: 321,169 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD05864766
Núm. CAS: 348640-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR