3-Bromo-o-anisaldehyde 95%

BLDPBD84958-5GEA 245 EUR
BLDPBD84958-5G BLDPBD84958-250MG BLDPBD84958-1G
3-Bromo-o-anisaldehyde 95%
3-Bromo-o-anisaldehyde

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₇BrO₂
MW: 215,0400 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD09834811
Núm. CAS: 88275-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR