3-Benzyloxybenzyl bromide 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD84737-10GEA 96 EUR
BLDPBD84737-10G BLDPBD84737-250MG BLDPBD84737-5G BLDPBD84737-25G BLDPBD84737-1G
3-Benzyloxybenzyl bromide 98%
3-Benzyloxybenzyl bromide

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₄H₁₃BrO
MW: 277,1600 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD01318307
Núm. CAS: 1700-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR