3',5'-Diacetoxyacetophenone 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD82354-25GEA 47 EUR
BLDPBD82354-25G BLDPBD82354-5G BLDPBD82354-100G BLDPBD82354-1G
3',5'-Diacetoxyacetophenone 98%
3',5'-Diacetoxyacetophenone

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₂H₁₂O₅
MW: 236,2207 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00008702
Núm. CAS: 35086-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR