4-(trans-4-Propylcyclohexyl)phenylboronic acid 95%

Proveedor: BLD PHARMATECH GMBH

BLDPBD8121-5GEA 40 EUR
BLDPBD8121-5G BLDPBD8121-25G BLDPBD8121-100G BLDPBD8121-1G BLDPBD8121-10G
4-(trans-4-Propylcyclohexyl)phenylboronic acid 95%
4-(trans-4-Propylcyclohexyl)phenylboronic acid

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₅H₂₃BO₂
MW: 246,1529 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD06658197
Núm. CAS: 146862-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR